Anti-ACTA2 Mouse Monoclonal Antibody
Catalog # ENZOENZ30931
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:Smooth Muscle Actin
- Antigen symbol:ACTA2
- Clonality:Monoclonal
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Form:Liquid, ready-to-use solution in PBS containing 0,1% sodium azide and 1% BSA
- Antigen synonyms:AAT6|alpha 2 actin|MYMY5|aortic smooth muscle|cell growth-inhibiting gene 46 protein|alpha-cardiac actin|alpha-actin-2|actin|GIG46|growth-inhibiting gene 46|ACTSA
- Purification:Affinity purified
- Pk:6 mL
Specifications
About this item
Host: Mouse, Isotype: IgG1
- For Immunohistochemistry, Western Blot
Recognizes human, monkey and rabbit actin isotypes alpha and gamma of skeletal, cardiac and smooth muscle.
Type: Primary
Antigen: ACTA2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human