Anti-Proteasome 19S Rpn10/S5A Subunit Mouse Monoclonal Antibody
Catalog # ENZOBMLPW92500025
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:Proteasome 19S Rpn10/S5A Subunit
- Antigen symbol:Proteasome 19S Rpn10/S5A
- Clonality:Monoclonal
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Western blot:Yes
- Seq:02
- Form:Liquid, in PBS containing 10 mM sodium azide
- Pk:25 µl
Specifications
About this item
Host: Mouse, Isotype: IgG1
- For Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes human proteasome 26S (19S regulator complex) (Rpn10/S5a subunit).
Type: Primary
Antigen: PROTEASOME 19S RPN10/S5A SUBUNIT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human