Order Entry
Export
ContactUsLinkComponent
Anti-BMF Rat Monoclonal Antibody
Anti-BMF Rat Monoclonal Antibody
Catalog # ENZOALX804342C100
Supplier:  ENZO LIFE SCIENCES
Anti-BMF Rat Monoclonal Antibody
Catalog # ENZOALX804342C100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Bcl2 modifying Factor
  • Antigen symbol:
    BMF
  • Clonality:
    Monoclonal
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 0,02% sodium azide
  • Concentration:
    1 mg/ml
  • Purification:
    Protein G-affinity purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Host: Rat, Isotype: IgG1

  • For Immunoprecipitation, Western Blot

Recognizes human and mouse Bmf.

Type: Primary
Antigen: BMF
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human