Order Entry
Export
ContactUsLinkComponent
Anti-RIG-I Rabbit Polyclonal Antibody
Anti-RIG-I Rabbit Polyclonal Antibody
Catalog # ENZOALX210932C100
Supplier:  ENZO LIFE SCIENCES
Anti-RIG-I Rabbit Polyclonal Antibody
Catalog # ENZOALX210932C100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Retinoic Acid-inducible Gene 1
  • Antigen symbol:
    RIG-1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 0,02% sodium azide.
  • Antigen synonyms:
    Probable ATP-dependent RNA helicase DDX58|Retinoic acid-inducible gene 1 protein|DEAD-box protein 58|RIGI|RIG-I|SGMRT2|RLR-1
  • Concentration:
    1 mg/ml
  • Purification:
    Protein A-affinity purified
  • Pk:
    100 µG

Specifications

About this item

RIG-I (retinoic acid-inducible gene I; Ddx58), a member of the DEAD-box RNA helicases, is activated by cytoplasmic dsRNA and 5’-ppp RNA produced during the viral replication. The protein is characterized by a N-terminal region with two caspase recruitment domains (CARD) and a C-terminal region harboring potential ATP-dependent RNA helicase activity. RIG-1 recruits the CARD adaptor inducing IFN-β (Cardif) in a CARD-CARD-dependent manner resulting in NF-κB and IRF3 activation.

  • For Immunoprecipitation, Western Blot

Recognizes human RIG-I.

Type: Primary
Antigen: RIG-I
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human