Order Entry
Export
ContactUsLinkComponent
Anti-AKT Rabbit Polyclonal Antibody
Anti-AKT Rabbit Polyclonal Antibody
Supplier:  ENZO LIFE SCIENCES
Certificates
Anti-AKT Rabbit Polyclonal Antibody
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Catalog No:
  • ENZOADIKAPPK004F
  • ENZOADIKAPPK004D
  • Antigen symbol:
  • AKT
  • AKT
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Reactivity:
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Form:
  • Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
  • Purification:
  • Protein A affinity purified

Specifications