Order Entry
Export
ContactUsLinkComponent
 
Anti-Histone H2AX Mouse Monoclonal Antibody
 
Anti-Histone H2AX Mouse Monoclonal Antibody

 

  • Catalog No:
  • ENZOADIKAMCC255F
  • ENZOADIKAMCC255D
  • Antigen name:
  • Histone H2AX
  • Histone H2AX
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Reactivity:
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • Form:
  • Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
  • Purification:
  • Protein G affinity purified