Order Entry
Export
ContactUsLinkComponent
Anti-TCP1A Rat Monoclonal Antibody
Anti-TCP1A Rat Monoclonal Antibody
Supplier:  ENZO LIFE SCIENCES
Certificates
Anti-TCP1A Rat Monoclonal Antibody
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Catalog No:
  • ENZOADICTA123F
  • ENZOADICTA123D
  • Antigen symbol:
  • TCP1A
  • TCP1A
  • Antigen name:
  • T-complex Protein 1 Subunit Alpha
  • T-complex Protein 1 Subunit Alpha
  • Antigen synonyms:
  • TRic|Tp63|c-cpn|Ccta|Cct1|p63|Tcp-1|CCT|AI528772
  • TRic|Tp63|c-cpn|Ccta|Cct1|p63|Tcp-1|CCT|AI528772
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rat
  • Rat
  • Isotype:
  • IgG2c
  • IgG2c
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Form:
  • Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
  • Purification:
  • Protein G affinity purified

Specifications