Anti-HGF Mouse Monoclonal Antibody
Catalog # ENZOADI905163100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:Hepatocyte growth factor
- Antigen symbol:HGF
- Clonality:Monoclonal
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS containing 0,01% sodium azide. Concentration 1 mg/ml.
- Antigen synonyms:Hepatocyte growth factor|HPTA|DFNB39|HGFB|Hepatopoeitin-A|Fibroblast-derived tumor cytotoxic factor|Lung fibroblast-derived mitogen|SF|F-TCF
- Purification:Protein G affinity purified
- Pk:100 µG
Specifications
About this item
HGF/SF (Hepatocyte growth factor/Scatter factor) is a multifunctional heterodimeric polypeptide. It mediates the growth and scattering of various cell types, epithelial mesenchymal transition, the formation of tubules and lumens, and promotes angiogenesis. The ligand-receptor pair has also shown to be uniquely involved in most human solid tumors and to participate in tumor development, invasion, and metastasis.
- For Immunoprecipitation, Western Blot
Recognizes human HGF/SF.
Type: Primary
Antigen: HGF
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human