Order Entry
Export
ContactUsLinkComponent
Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate ≥97%
Irritant
Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate ≥97%
Catalog # SIAL754730-1G
Supplier:  Merck
CAS Number:  
Irritant
Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate ≥97%
Catalog # SIAL754730-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Colour:
    Orange to very dark orange, red to very dark red, orange-red and red-orange
  • Form:
    Powder or crystal or chunk
  • Phosphorus NMR spectra :
    Conforms to structure
  • Proton NMR spectrum:
    Conforms to structure
  • Fluorine NMR spectra:
    Conforms to structure
  • Carbon (C):
    40,5 – 43,5%
  • Nitrogen (N):
    9,4 – 10,1%
  • Assay:
    97%
  • Absorption:
    291 nm 80000 and 456 nm 13400

Specification Test Results

About this item

 :
Melting Pt:>300 °C
 :859.562 g/mol
 :GHS07
CAS Number:60804-74-2
Formula:C₃₀H₂₄F₁₂N₆P₂Ru
MDL Number:MFCD11042502