Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Density Standards
Density standards, ultra pure water
Density standards, ultra pure water
PAAR96044
:
Anton Paar
:
Density standards, ultra pure water
PAAR96044
:
Anton Paar
:
Pk:
1
Description:
Density standard, ultra-pure water, 5×10 ml, with certificate