Order Entry
Export
ContactUsLinkComponent
 
Syringe filters, MN Steriliser CA
  MANA740401.50
undefined
Syringe filters, MN Steriliser CA
  MANA740401.50

 

  • Ø:
    28 mm
  • Filtration area:
    6,2 cm²
  • Membrane:
    CA
  • Pore size:
    0,20 µm
  • Sterility:
    Sterile
  • Max. temperature:
    50 °C
  • Description:
    MN steriliser CA
  • Hold-up volume:
    150 µl
  • Max. operating pressure:
    4,5 bar
  • Pk:
    1
  • Inlet/outlet connections:
    Female Luer lock/male slip Luer

 

 

MN steriliser CA syringe filter units are designed to sterilise and remove microorganisms and particulates from aqueous solutions such as biological fluids, culture media, additives, buffers, and protein solutions.

  • Low protein binding
  • Very robust membrane - high stability
  • Fast flow of liquid through the filter
  • Pre-sterilised with ethylene oxide

Housing material: Methyl methacrylate-butadiene-styrene polymer (MBS)