Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D-Xylulose
D-Xylulose ≥95% in aqueous solution
D-Xylulose ≥95% in aqueous solution
D-Xylulose ≥95% in aqueous solution
Supplier:
Cayman Chemical
Certificates
Expand image
D-Xylulose ≥95% in aqueous solution
Supplier:
Cayman Chemical
About this item
CAS Number:
551-84-8
:
Formula:
C₅H₁₀O₅
MDL Number:
MFCD00065958
:
150.13 g/mol
Storage Temperature:
Freezer