Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Rivastigmine-L-tartrate
Rivastigmine-L-tartrate ≥95%, crystalline solid
Rivastigmine-L-tartrate ≥95%, crystalline solid
Toxic
Health
Environment
Rivastigmine-L-tartrate ≥95%, crystalline solid
Supplier:
Cayman Chemical
Certificates
Expand image
Toxic
Health
Environment
Rivastigmine-L-tartrate ≥95%, crystalline solid
Supplier:
Cayman Chemical
About this item
CAS Number:
129101-54-8
:
Formula:
C₁₄H₂₂N₂O₂·C₄H₆O₆
MDL Number:
MFCD03700731
:
400.43 g/mol
:
GHS06
:
GHS08
:
GHS09
Storage Temperature:
Freezer