Shoe covers, 402
Blue, 150 mm, 250 - 380 mm
Catalog # MMMA402
Supplier: 3M
Jump to:
Product Details & Documents
These disposable shoe covers made from now woven PP are designed to provide basic barrier protection to help protect footwear against light liquid splashes and hazardous dusts.
- Elastic top and bottom for protection against contaminants and enhanced fit
- Sole and upper constructed of the same material
- Anti-static treated
Weight: 40 g/m²
Length: 250 - 380 mm
Height: 150 mm
Opening diameter: 1,5 - 15 cm
Documents
TDS Overshoe cover 402 blue
Datasheet
Specifications
- Colour:Blue
- Height:150 mm
- Length:250 - 380 mm
- Cat. No.:MMMA402
- Anti-static:+
- Garment application:General Purpose|Medical|Laboratory|Industrial|Food
- Fabric:PP
- Weight:40 g/m²



