Leather gloves, A271
Supplier: Portwest
About this item
Premium quality leather drivers glove with Insulatex™ lining for added warmth and comfort.
- Made from cow grain leather
- Provide mechanical and cold protection
: EN 388:2016 (3142X), EN 511 (22X)
2 Options Available Below
Product Details & Documents
Premium quality leather drivers glove with Insulatex™ lining for added warmth and comfort.
- Made from cow grain leather
- Provide mechanical and cold protection
: EN 388:2016 (3142X), EN 511 (22X)
Specifications for Product Family
Specifications
- Catalog No:
- PRTWA271TARL
- PRTWA271TARXL
- Model:
- A271
- A271
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Liner material:
- Insulatex™
- Insulatex™
- EN norm:
- EN 388:2016 (3142X), EN 511 (22X)
- EN 388:2016 (3142X), EN 511 (22X)
Specifications
Product Family Options
Product Information
- Glove sizeColourAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.PRTWA271TARLModelA271Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLiner materialInsulatex™EN normEN 388:2016 (3142X), EN 511 (22X) - Specifications:
More
Specifications
Cat. No.PRTWA271TARXLModelA271Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLiner materialInsulatex™EN normEN 388:2016 (3142X), EN 511 (22X)