Anti-SP1 Polyclonal Antibody
Catalog # BSBTRP1072
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:SP1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for SP1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. SP1 information: Molecular Weight: 80693 MW; Subcellular Localization: Nucleus. Cytoplasm. Nuclear location is governed by glycosylated/phosphorylated states. Insulin promotes nuclear location, while glucagon favors cytoplasmic location; Tissue Specificity: Up-regulated in adenocarcinomas of the stomach (at protein level). Isoform 3 is ubiquitously expressed at low levels.
Type: Primary
Antigen: SP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human