Anti-Pin1 Polyclonal Antibody
BSBTPB9316
- Antibody type:Primary
- Antigen symbol:Pin1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Polyclonal antibody for PIN1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. PIN1 information: Molecular Weight: 18243 MW; Subcellular Localization: Nucleus . Nucleus speckle . Cytoplasm . Colocalizes with NEK6 in the nucleus (PubMed:16476580). Mainly localized in the nucleus but phosphorylation at Ser-71 by DAPK1 results in inhibition of its nuclear localization (PubMed:21497122); Tissue Specificity: The phosphorylated form at Ser-71 is expressed in normal breast tissue cells but not in breast cancer cells.
Type: Primary
Antigen: Pin1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat