Anti-HNF-4-Alpha Polyclonal Antibody
Catalog # BSBTPB9215
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:HNF-4-Alpha
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for NR2A1/HNF4A detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. NR2A1/HNF4A information: Molecular Weight: 52785 MW; Subcellular Localization: Nucleus.
Type: Primary
Antigen: HNF-4-Alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat