Order Entry
Export
ContactUsLinkComponent
 
Anti-FGFR4 Polyclonal Antibody
  BSBTPB9193
undefined
Anti-FGFR4 Polyclonal Antibody
  BSBTPB9193

 

  • Antibody type:
    Primary
  • Antigen symbol:
    FGFR4
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Polyclonal antibody for FGFR 4/FGFR4 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. FGFR 4/FGFR4 information: Molecular Weight: 87954 MW; Subcellular Localization: Cell membrane; Single-pass type I membrane protein. Endosome. Endoplasmic reticulum. Internalized from the cell membrane to recycling endosomes, and from there back to the cell membrane; Tissue Specificity: Expressed in gastrointestinal epithelial cells, pancreas, and gastric and pancreatic cancer cell lines.

Type: Primary
Antigen: FGFR4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human