Order Entry
Export
ContactUsLinkComponent
Anti-CBFb Polyclonal Antibody
Anti-CBFb Polyclonal Antibody
Catalog # BSBTA01007-1
Supplier:  Boster Bio
undefined
Anti-CBFb Polyclonal Antibody
Catalog # BSBTA01007-1
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CBFb
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for CBFb detection. Tested with WB, IHC-P, Direct ELISA in Human;Mouse;Rat.

Type: Primary
Antigen: CBFb
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat