Anti-Adiponectin Polyclonal Antibody
Catalog # BSBTPB9001
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Adiponectin
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for Adiponectin/ADIPOQ detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. Adiponectin/ADIPOQ information: Molecular Weight: 26414 MW; Subcellular Localization: Secreted; Tissue Specificity: Synthesized exclusively by adipocytes and secreted into plasma.
Type: Primary
Antigen: Adiponectin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human