
Guanidinium hydrochloride ≥99%, white crystalline powder for molecular biology
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:50-01-1
:
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026
:95.53 g/mol
Melting Pt:180…190 °C
Density:1.345 g/cm³ (20 °C) (lit.)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
- Melting Point:180 - 190°C
- FTIR:Conforms to Standard
- pH (6.0 M aq soln):4-6
- OD260 (6.0 M aq soln):≤0.03
- Loss on Drying:≤1.0%
- Solubility (6.0 M aq soln):Clear, Colorless Solution