Glycine ≥99% for molecular biology
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH
CAS Number:56-40-6
:
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131
Boiling Pt:181
Melting Pt:240 °C (dec., lit.)
Density:1.16 g/mL (lit.)
Molecular Weight:75.07 g/mol
Merck Index:13,04504
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
- FTIR:Conforms to Standard
- Loss on Drying:≤0.5%