
Sertraline hydrochloride
:79559-97-0
:
:C₁₇H₁₇Cl₂N
:MFCD00895772
:342.69 g/mol
:245 °C
:GHS07
:Freezer
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Water:≤0.5 %
- HPLC:≥97.5 %
- Specific optical rotation:+38° to +43° (20°C, 589 nm) (2 % in CH3OH)