2,4-Dichloroquinazoline 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:607-68-1
:
Formula:C₈H₄Cl₂N₂
MDL Number:MFCD00091950
:199.04 g/mol
Melting Pt:115…121 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow to light brown Powder or crystalline powder
- Infrared spectrum:Conforms
- Melting point:116 to 123 °C
- GC:≥97.5 %