Order Entry
Export
Orders LinkContactUsLinkComponent
Estriol 97%

Estriol 97%

Health
Estriol 97%
Certificates
Estriol 97%

2 Options Available Below

Product Details & Documents
  • CAS Number:50-27-1
  • Formula:C₁₈H₂₄O₃
  • MDL Number:MFCD00003691
  • Melting Pt:280…282 °C
  • Storage Temperature:Ambient
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 %
  • HPLC:
    ≥96.0 %
  • Specific optical rotation:
    +54° to +62° (20°C, 589 nm) (0.4 % in Dioxane)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 101,00 €
    101,00 €
    1 * 1 g
       101,00 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 265,00 €
    265,00 €
    1 * 5 g
       265,00 €