1,6-Naphthyridine 96%
1 g
Catalog # ACRO457670010
Supplier: Thermo Fisher Scientific
1,6-Naphthyridine 96%
Catalog # ACRO457670010
Supplier: Thermo Fisher Scientific
CAS Number: 253-72-5
Jump to:
Product Details & Documents
Synonyms: Pyrido[4,3-b]pyridine, 1,6-Diazanaphthalene
- CAS Number:253-72-5
- Formula:C₈H₆N₂
- MDL Number:MFCD00059750
- Molecular Weight:130.15 g/mol
- Melting Pt:36…38 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:Off-white to brown Low melting solid
- Infrared spectrum:Conforms
- GC:≥95.0 %
Specifications
- Pk:1 g



