
2,2'-Bis(trifluoromethyl)benzidine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:341-58-2
:
Formula:C₁₄H₁₀F₆N₂
MDL Number:MFCD00190155
:320.24 g/mol
Melting Pt:183…184 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to off-white to light brown Powder
- Infrared spectrum:Conforms
- Melting point:180 to 186 °C
- HPLC:≥96.0 %