1,3-Benzenedithiol 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:626-04-0
:
Formula:C₆H₆S₂
MDL Number:MFCD00004842
:142.25 g/mol
Boiling Pt:128…129 °C (21hPa)
Melting Pt:24 °C
Density:1.24 g/cm³
Flash Pt:113 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to white to light yellow Liquid or solid
- Infrared spectrum:Conforms
- GC:≥94 %