Order Entry
Export
ContactUsLinkComponent
 
 
1,10-Phenanthroline-5,6-dione 98%
 
undefined
 
1,10-Phenanthroline-5,6-dione 98%

 

  • Appearance:
    Yellow to orange Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    255 to 260 °C
  • HPLC:
    ≥96.0 %