
1,4-Cineole 85%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:470-67-7
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00209502
:154.25 g/mol
Boiling Pt:65 °C (21hPa)
Melting Pt:–46 °C
Density:0.887 g/cm³
Flash Pt:47 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4450 to 1.4560 (20°C, 589 nm)
- GC:≥80 %