
Imidazo[1,2-b]pyridazine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:766-55-2
:
Formula:C₆H₅N₃
MDL Number:MFCD07782103
:119.12 g/mol
Melting Pt:54 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to light grey Solid
- Infrared spectrum:Conforms
- Melting point:50 to 57 °C
- Water:≤3 %
- GC:≥97.5 %