
2,4-Dihydroxy-6-methylbenzoic acid hydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:480-64-8
:
Formula:C₈H₈O₄·xH₂O
MDL Number:MFCD00210536
:168.15 g/mol (anhy)
Melting Pt:152…153 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow Powder or solid
- Infrared spectrum:Conforms
- Water:≤10 %
- HPLC:≥97.5 %