Order Entry
Export
Orders LinkContactUsLinkComponent
 

Quartz, fine wool 4 µm

Health
Quartz, fine wool 4 µm
Certificates
Quartz, fine wool 4 µm

2 Options Available Below

Product Details & Documents
Synonyms:  Silicium dioxide, Silicium (IV) oxide, Silicon dioxide, Sand, white quartz
  • CAS Number:14808-60-7
  • Formula:SiO₂
  • MDL Number:MFCD00217788
  • Molecular Weight:60.08 g/mol
  • Storage Temperature:Ambient
Specifications for Product Family

Specification Test Results

  • Appearance:
    White Fibers
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 51,00 €
    51,00 €
    1 * 10 g
       51,00 €

  • Glass bottle
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 181,00 €
    181,00 €
    1 * 50 g
       181,00 €