
4,4'-Dimethyldiphenylamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:620-93-9
:
Formula:C₁₄H₁₅N
MDL Number:MFCD00059317
:197.2756 g/mol
Melting Pt:80…82 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light brown Powder
- Infrared spectrum:Conforms
- Melting point:77 to 84 °C
- HPLC:≥96.0 %