

10-Deacetylbaccatin III 99%
ACRO294770010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White Powder
- Infrared spectrum:Conforms
- HPLC:≥98.5 %
- Specific optical rotation:-43° to -46° (25°C, 589 nm) (c=1, CH3OH)
:
:221…223 °C
:544.60 g/mol
:GHS07
:GHS08
:Ambient
:32981-86-5
:C₂₉H₃₆O₁₀
:MFCD00132913