Order Entry
Export
Orders LinkContactUsLinkComponent
2,2':6',2''-Terpyridine 96%

2,2':6',2''-Terpyridine 96%

Toxic
2,2':6',2''-Terpyridine 96%
Certificates
2,2':6',2''-Terpyridine 96%

3 Options Available Below

Product Details & Documents
Synonyms:  TERPY
  • CAS Number:1148-79-4
  • Formula:C₁₅H₁₁N₃
  • MDL Number:MFCD00006213
  • Molecular Weight:233.27 g/mol
  • Boiling Pt:370 °C
  • Melting Pt:89…91 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,I
Specifications for Product Family

Specification Test Results

  • Appearance:
    Cream to yellow to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    89 to 95 °C
  • GC:
    ≥95 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 52,50 €
    52,50 €
    1 * 250 mg
       52,50 €

  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 164,00 €
    164,00 €
    1 * 1 g
       164,00 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 540,00 €
    540,00 €
    1 * 5 g
       540,00 €