Order Entry
Export
ContactUsLinkComponent
Sleeve protector, Tychem® F
Sleeve protector, Tychem® F
  SSSAAS-TF-02DP
undefined
Sleeve protector, Tychem® F
  SSSAAS-TF-02DP

 

  • Garment application:
    Laboratory
  • Colour:
    Grey
  • Length:
    500 mm
  • Seq:
    01
  • Pk:
    50

 

 

These sleeve protectors are made from chemical resistant Tychem® F fabric.

  • Elasticated opening
  • Tight seems

Size: 47 cm

 : Cat. I