Aztreonam, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1046205.
- 1046307.
- 1046409.
- Description:
- Aztreonam
- Aztreonam E-Isomer
- Open Ring Aztreonam
- Synonym(s):
- (2S,3S)-2-{(Z)-2-[2-Aminothiazol-4-yl]-2-[2-carboxypropan-2-yloxyimino]acetamido}-3-(sulfoamino)butanoic acid
- CAS No.:
- 78110-38-0
- 87500-74-1
- Formula:
- C₁₃H₁₇N₅O₈S₂
- C₁₃H₁₇N₅O₈S₂
- C₁₃H₁₉N₅O₉S₂
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1046205.AztreonamSpecifications:
More
Specifications
Cat. No.1046205.DescriptionAztreonamSynonym(s)CAS No.78110-38-0FormulaC₁₃H₁₇N₅O₈S₂ - 1046307.Aztreonam E-IsomerSpecifications:
More
Specifications
Cat. No.1046307.DescriptionAztreonam E-IsomerSynonym(s)CAS No.FormulaC₁₃H₁₇N₅O₈S₂ - 1046409.Open Ring AztreonamSpecifications:
More
Specifications
Cat. No.1046409.DescriptionOpen Ring AztreonamSynonym(s)(2S,3S)-2-{(Z)-2-[2-Aminothiazol-4-yl]-2-[2-carboxypropan-2-yloxyimino]acetamido}-3-(sulfoamino)butanoic acidCAS No.87500-74-1FormulaC₁₃H₁₉N₅O₉S₂



