Lubricants, APIEZON®
Supplier: Roth Carl
Certificates
About this item
High lubricating and gettering properties. Extremely low steam pressures.
- Chemically inert
- Silicone-free
- Non toxic
- Temperature resistant
- 10 years shelf life
Specifications
- Catalog No:
- ROTHC986.1
- ROTHC987.1
- ROTH1683.1
- ROTH1686.1
- Temperature range:
- 10...30 °C
- −40...+180 °C
- −10...+240 °C
- Type:
- Apiezon® AP100
- Apiezon® AP101
- Apiezon® H
- Vapor pressure:
- 7 x 10-11 Torr (20 °C)
- <1.0 x 10-5 Torr (20 °C)
- 1,7 x 10-9 Torr (20 °C)