Order Entry
Export
ContactUsLinkComponent
1,4-Bis(3-aminopropyl)piperazine ≥99%, Sigma-Aldrich®
1,4-Bis(3-aminopropyl)piperazine ≥99%, Sigma-Aldrich®
Catalog # SIAL239488-50ML
Supplier:  Merck
CAS Number:  
1,4-Bis(3-aminopropyl)piperazine ≥99%, Sigma-Aldrich®
Catalog # SIAL239488-50ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • CAS Number:
    7209-38-3
  • Empirical Formula (Hill Notation):
    C₁₀H₂₄N₄
  • Molecular Weight:
    200,32
  • EC Number:
    230-589-1
  • MDL number:
    MFCD00006161
  • PubChem Substance ID:
    24854378
  • Vapor density:
    6,9 (vs air)
  • InChI Key:
    XUSNPFGLKGCWGN-UHFFFAOYSA-N
  • Assay:
    ≥99%
  • Refractive index:
    n20/D 1,502(lit.)
  • Bp:
    150 - 152 °C/2 mm Hg(lit.)
  • Mp:
    15 °C(lit.)
  • Density:
    0,973 g/ml at 25 °C(lit.)
  • Gene Information:
    rat ... Grin2a(24409)

Specification Test Results

About this item

 :
 :200.33 g/mol
Density:0.973 g/ml (25 °C) (lit.)
 :M-Clarity™ quality level = MQ100
CAS Number:7209-38-3
Formula:C₁₀H₂₄N₄
MDL Number:MFCD00006161