Order Entry
Export
ContactUsLinkComponent
Organic reference standard, PAH Mix 61 in Methanol/Acetone 1:1, Dr. Ehrenstorfer
Organic reference standard, PAH Mix 61 in Methanol/Acetone 1:1, Dr. Ehrenstorfer
Catalog # EHERXA06100100AM
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, PAH Mix 61 in Methanol/Acetone 1:1, Dr. Ehrenstorfer
Catalog # EHERXA06100100AM
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 mL
  • Standards:
    Reference standard
  • Description:
    PAH Mix 61 in Methanol/Acetone 1:1
  • Description:
    PAH Mix 61 in Methanol/Acetone 1:1

Specifications

About this item

  • Food and environment industry applications