Enoxaparin Sodium, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1235820.
- 1235831.
- Description:
- Enoxaparin Sodium
- Enoxaparin Sodium for Bioassays
- CAS No.:
- 679809-58-6
- 9041-08-1
- Formula:
- (C₁₂H₁₆NS₂Na₃)₂₀
- (C₁₂H₁₆NS₂Na₃)₂₀
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1235820.Enoxaparin SodiumSpecifications:
More
Specifications
Cat. No.1235820.DescriptionEnoxaparin SodiumCAS No.679809-58-6Formula(C₁₂H₁₆NS₂Na₃)₂₀ - 1235831.Enoxaparin Sodium for BioassaysEnoxaparin Sodium for BioassaysUSP reference standardAmpule1Log in for availability1 * 1 items875,00 €Specifications:
More
Specifications
Cat. No.1235831.DescriptionEnoxaparin Sodium for BioassaysCAS No.9041-08-1Formula(C₁₂H₁₆NS₂Na₃)₂₀



