Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Food Standards
Aminomethyl phosphonicacid N-acetyl, Dr. Ehrenstorfer
Aminomethyl phosphonicacid N-acetyl, Dr. Ehrenstorfer
Aminomethyl phosphonicacid N-acetyl, Dr. Ehrenstorfer
Catalog # EHERC10205150
Supplier:
EHRENSTORFER
CAS Number:
Expand image
Aminomethyl phosphonicacid N-acetyl, Dr. Ehrenstorfer
Catalog # EHERC10205150
Supplier:
EHRENSTORFER
CAS Number:
Specifications
Pk:
10 mg
Description:
Aminomethyl phosphonic acid N-acetyl, reference standard for food analysis
Specifications
About this item
Product Details
Product Details