Order Entry
Export
Orders LinkContactUsLinkComponent
Mirtazapine, Reference Standards

Mirtazapine, Reference Standards

Supplier:   USP
Mirtazapine, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Mirtazapine Resolution Mixture: This mixture contains mirtazapine, impurity A, impurity B, impurity C, impurity D, impurity E and impurity F

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1444279.
  • 1444296.
  • Description:
  • Mirtazapine
  • Mirtazapine Resolution Mixture
  • CAS No.:
  • 85650-52-8
  • Formula:
  • C₁₇H₁₉N₃

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1444279.
    Mirtazapine
    Mirtazapine
    USP reference standard
    Vial
    350 mg
    Log in for availability
    Log in for availability
    1 * 350 mg
    1 * 350 mg - 1 780,00 €
    1 780,00 €
    1 * 350 mg
       1 780,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1444279.
    Description
    Mirtazapine
    CAS No.
    85650-52-8
    Formula
    C₁₇H₁₉N₃
  • 1444296.
    Mirtazapine Resolution Mixture
    Mirtazapine Resolution Mixture
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 640,00 €
    1 640,00 €
    1 * 25 mg
       1 640,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1444296.
    Description
    Mirtazapine Resolution Mixture
    CAS No.
    Formula