Order Entry
Export
Orders LinkContactUsLinkComponent
 

Clobetasol Propionate, Reference Standards

 :   USP
Clobetasol Propionate, Reference Standards
 :  USP
 
Clobetasol Propionate, Reference Standards

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

 

 

 

  • Catalog No:
  • 1138405.
  • 1138427.
  • Description:
  • Clobetasol Propionate
  • Clobetasol Propionate Related Compound A
  • Synonym(s):
  • 9-α-fluoro-11-β-hydroxy-16-β-methyl-3-oxoandrosta-1,4-diene-17(R)-spiro-2'-[4'-chloro-5'-ethylfuran-3'(2'H)-one]
  • CAS No.:
  • 25122-46-7
  • 1486466-31-2
  • Formula:
  • C₂₅H₃₂ClFO₅
  • C₂₅H₃₀ClFO₄

 

Product Family Options

 

  • Description
    Standards
    Pack type
    Pk
     
     
  • Clobetasol Propionate
    USP reference standard
    Vial
    200 mg
       
       
    1 * 200 mg
    1 * 200 mg - 406,00 €
    406,00 €
    1 * 200 mg
       406,00 €

     
     
    Specifications
    Cat. No.
    1138405.
    Description
    Clobetasol Propionate
    Synonym(s)
    CAS No.
    25122-46-7
    Formula
    C₂₅H₃₂ClFO₅
  • Clobetasol Propionate Related Compound A
    USP reference standard
    Vial
    50 mg
       
       
    1 * 50 mg
    1 * 50 mg - 1 230,00 €
    1 230,00 €
    1 * 50 mg
       1 230,00 €

     
     
    Specifications
    Cat. No.
    1138427.
    Description
    Clobetasol Propionate Related Compound A
    Synonym(s)
    9-α-fluoro-11-β-hydroxy-16-β-methyl-3-oxoandrosta-1,4-diene-17(R)-spiro-2'-[4'-chloro-5'-ethylfuran-3'(2'H)-one]
    CAS No.
    1486466-31-2
    Formula
    C₂₅H₃₀ClFO₄