Order Entry
Export
Orders LinkContactUsLinkComponent
Fluticasone Propionate, Reference Standards

Fluticasone Propionate, Reference Standards

Supplier:   USP
Fluticasone Propionate, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1285895.
  • 1285910.
  • 1285873.
  • Description:
  • Fluticasone Propionate System Suitability Mixture
  • Fluticasone Propionate Nasal Spray Resolution Mixture
  • Fluticasone Propionate
  • Synonym(s):
  • Fluticasone Propionate and Fluticasone Propionate Related Compounds B, C and D
  • CAS No.:
  • 80474-14-2
  • Formula:
  • C₂₅H₃₁F₃O₅S

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1285895.
    Fluticasone Propionate System Suitability Mixture
    Fluticasone Propionate System Suitability Mixture
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 640,00 €
    1 640,00 €
    1 * 25 mg
       1 640,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1285895.
    Description
    Fluticasone Propionate System Suitability Mixture
    Synonym(s)
    Fluticasone Propionate and Fluticasone Propionate Related Compounds B, C and D
    CAS No.
    Formula
  • 1285910.
    Fluticasone Propionate Nasal Spray Resolution Mixture
    Fluticasone Propionate Nasal Spray Resolution Mixture
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 670,00 €
    1 670,00 €
    1 * 25 mg
       1 670,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1285910.
    Description
    Fluticasone Propionate Nasal Spray Resolution Mixture
    Synonym(s)
    CAS No.
    Formula
  • 1285873.
    Fluticasone Propionate
    Fluticasone Propionate
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 1 760,00 €
    1 760,00 €
    1 * 100 mg
       1 760,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1285873.
    Description
    Fluticasone Propionate
    Synonym(s)
    CAS No.
    80474-14-2
    Formula
    C₂₅H₃₁F₃O₅S