Order Entry
Export
ContactUsLinkComponent
 
Human recombinant DcR3 (from CHO cells)
Catalog # PRSI90-454
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant DcR3 (from CHO cells)
Catalog # PRSI90-454
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein synonyms:
    Tumor Necrosis Factor Receptor Superfamily Member 6B|TNFRSF6B|Decoy Receptor 3|M68
  • Protein/peptide name:
    DcR3
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    24-300
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.

Specifications

About this item

Decoy receptor 3 (DcR3; TNFRSF6B; M68) is a member of the TNF receptor superfamily. DcR3 shares sequence identity with OPG (31%), TNF-RII (29%) and Fas (17%) and is expressed in a variety of different tissues and at high levels in many malignant tumours. Ligands of DcR3 include Fas ligand, homologous to lymphotoxin showing inducible expression and competing with HSV glycoprotein D for herpes virus entry mediator (a receptor expressed by T lymphocytes), TNF-like molecule 1A and heparan sulfate proteoglycans. DcR3 modulates the function of T cells, dendritic cells and macrophages. DcR3/Fc fusion proteins can bind to human and mouse B cells and suppress the activation of B cells.

Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.