Order Entry
Export
Orders LinkContactUsLinkComponent
Human recombinant BMP2 (from <i>E. coli</i>)

Human recombinant BMP2 (from E. coli)

10 µG
Catalog # PRSI90-373
Supplier:   ProSci Inc.
Human recombinant BMP2 (from <i>E. coli</i>)
Catalog # PRSI90-373
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant BMP2 (from E. coli)

10 µG

CAS Number:  
Price
364,00 €
Log in for availability
1 * 10 µG
Product Details & Documents

Bone morphogenetic protein 2 (BMP-2) is a member of the BMP subgroup of the TGF-beta superfamily. It plays a dominant role in embryonic dorsalventral patterning, organogenesis, limb bud formation and bone formation and regeneration. BMP-2 signals through heterodimeric complexes composed of a type I receptor (Activin RI, BMP-RIA or BMP-RIB) and a type II receptor (BMP-RII or Activin RIIB). BMP-2 induces chondrocyte proliferation, endochondral bone formation, longitudinal bone growth and bone and cartilage repair. It induces ectopic bone formation or calcification by promoting osteogenic and chondrogenic differentiation in mesenchymal cells, stem cells and vascular smooth muscle cells. It also promotes the maintenance and repair of colonic epithelium, suppresses neuronal dopamine synthesis and release, induces apoptosis in medulloblastoma cells and is required for cardiac contractility.

Fusion-Tag: His Tag

This recombinant proteins is for research use only.

Documents

Specifications

Specifications

  • Pk:
    10 µG
  • Cat. No.:
    PRSI90-373
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    BMP2
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.
  • Protein synonyms:
    Bone Morphogenetic Protein 2|BMP2
  • Purity:
    >95% (SDS-PAGE)
  • Tag sequence:
    His Tag
  • Sequence:
    289-396
  • Species:
    Human
  • Source:
    E. coli

Specifications