Human recombinant LIGHT (soluble) (from CHO cells)
10 µG
Catalog # PRSI90-224
Supplier: ProSci Inc.
Human recombinant LIGHT (soluble) (from CHO cells)
Catalog # PRSI90-224
Supplier: ProSci Inc.
CAS Number:
Jump to:
Product Details & Documents
LIGHT (HVEML; CD258) is a cytokine that binds to TNFRSF3/LTBR. It activates NF-kappaB, stimulates the proliferation of T cells, and inhibits growth of the adenocarcinoma HT-29. Acts as a receptor for Herpes simplex virus.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.
Documents
75793-954.pdf
Datasheet
Specifications
Specifications
- Pk:10 µG
- Cat. No.:PRSI90-224
- Protein/peptide type:Recombinant
- Protein/peptide name:LIGHT (soluble)
- Storage conditions:Stable for at least 6 months after receipt when stored at −20 °C.
- Formulation:Lyophilized, Contains PBS.
- Protein synonyms:CD258|Tumor Necrosis Factor Ligand Superfamily Member 14|HVEML|TNFSF14
- Purity:>95% (SDS-PAGE)
- Tag sequence:DDDDK Tag
- Sequence:89-240
- Species:Human
- Source:CHO cells



